Product Details

SNP ID
rs73015352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170715688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGATGGGTGCAGCTGGAGCCAGGC[A/C]TAGGGGTGCCCACGCCAACTTCACA
Phenotype
MIM: 609391
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01124 PubMed Links

Gene Details

Gene
LINC01124
Gene Name
long intergenic non-protein coding RNA 1124
There are no transcripts associated with this gene.

Gene
LOC101926913
Gene Name
uncharacterized LOC101926913
There are no transcripts associated with this gene.

Gene
SP5
Gene Name
Sp5 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003845.2 964 Intron NP_001003845.1
XM_005246542.4 964 UTR 5 XP_005246599.1
XM_011511159.2 964 UTR 5 XP_011509461.1

View Full Product Details