Product Details

SNP ID
rs73975907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:173906814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGCAGCTGTTTCAAAACACATA[C/T]ACACTCCATCTCCAGAGATGTTGAT
Phenotype
MIM: 601804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SP3 PubMed Links

Gene Details

Gene
SP3
Gene Name
Sp3 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017371.4 5995 UTR 3 NP_001017371.3
NM_001172712.1 5995 UTR 3 NP_001166183.1
NM_003111.4 5995 UTR 3 NP_003102.1

View Full Product Details