Product Details

SNP ID
rs73977802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182836076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTAATTCTTCATTTCAGTATT[C/T]TTTACTCCACAATCTTGTCTTCATA
Phenotype
MIM: 605083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRZB PubMed Links

Gene Details

Gene
FRZB
Gene Name
frizzled-related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001463.3 Intron NP_001454.2

View Full Product Details