Product Details

SNP ID
rs73074987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219180239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCAGTACAAGAGGCAGAATCC[A/G]GCTCAGGTAACCTCCCCTACATCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321389.1 712 Intron NP_001308318.1
NM_001321390.1 712 Intron NP_001308319.1
NM_001321391.1 712 Intron NP_001308320.1
NM_015680.5 712 Intron NP_056495.3
Gene
FAM134A
Gene Name
family with sequence similarity 134 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321109.1 712 UTR 5 NP_001308038.1
NM_001321110.1 712 UTR 5 NP_001308039.1
NM_024293.5 712 Silent Mutation CCA,CCG P183P NP_077269.3
XM_005246848.2 712 Silent Mutation CCA,CCG P183P XP_005246905.1

View Full Product Details