Product Details

SNP ID
rs72995155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:158799407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAAATGGATCGATTTGGATACAG[C/T]GAAACTTTGAAAAGGTCTCTTGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAPL1 PubMed Links

Gene Details

Gene
DAPL1
Gene Name
death associated protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017920.2 Intron NP_001017920.2

View Full Product Details