Product Details

SNP ID
rs67810343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241011592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATCCTTTCCCCTTGTTTTCCAC[A/G]TCTCTATGACATGTTTTACAAGGTG
Phenotype
MIM: 616634
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNED1 PubMed Links

Gene Details

Gene
SNED1
Gene Name
sushi, nidogen and EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080437.1 Intron NP_001073906.1
XM_011510931.2 Intron XP_011509233.1
XM_011510932.2 Intron XP_011509234.1
XM_011510933.2 Intron XP_011509235.1
XM_011510934.2 Intron XP_011509236.1
XM_011510937.2 Intron XP_011509239.1
XM_017003773.1 Intron XP_016859262.1
XM_017003774.1 Intron XP_016859263.1
XM_017003775.1 Intron XP_016859264.1
XM_017003776.1 Intron XP_016859265.1
XM_017003777.1 Intron XP_016859266.1
XM_017003778.1 Intron XP_016859267.1
XM_017003779.1 Intron XP_016859268.1
XM_017003780.1 Intron XP_016859269.1
XM_017003781.1 Intron XP_016859270.1
XM_017003782.1 Intron XP_016859271.1

View Full Product Details