Product Details

SNP ID
rs73011977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241797375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCGTGACTGTTTAGAATTTTTC[C/T]GGCCATCACTATATGTTTTTTGTCC
Phenotype
MIM: 608237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST2 PubMed Links

Gene Details

Gene
GAL3ST2
Gene Name
galactose-3-O-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022134.2 Intron NP_071417.2

View Full Product Details