Product Details

SNP ID
rs73104075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240470764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGATCACAGCAACACAGTAGA[C/T]TCAGCTAAAATAACCCCCTCTCCTC
Phenotype
MIM: 600395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKMY1 PubMed Links

Gene Details

Gene
ANKMY1
Gene Name
ankyrin repeat and MYND domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282771.1 Intron NP_001269700.1
NM_001282780.1 Intron NP_001269709.1
NM_001282781.1 Intron NP_001269710.1
NM_001308375.1 Intron NP_001295304.1
NM_016552.3 Intron NP_057636.2
NM_017844.3 Intron NP_060314.2
XM_011511300.2 Intron XP_011509602.1
XM_011511301.1 Intron XP_011509603.1
XM_011511304.2 Intron XP_011509606.1
XM_011511305.2 Intron XP_011509607.1
XM_011511309.2 Intron XP_011509611.1
XM_011511313.2 Intron XP_011509615.1
XM_011511315.2 Intron XP_011509617.1
XM_011511317.1 Intron XP_011509619.1
XM_011511318.1 Intron XP_011509620.1
XM_011511319.2 Intron XP_011509621.1
XM_011511322.1 Intron XP_011509624.1
XM_017004271.1 Intron XP_016859760.1
XM_017004272.1 Intron XP_016859761.1
XM_017004273.1 Intron XP_016859762.1
XM_017004274.1 Intron XP_016859763.1
XM_017004275.1 Intron XP_016859764.1
XM_017004276.1 Intron XP_016859765.1
XM_017004277.1 Intron XP_016859766.1
XM_017004278.1 Intron XP_016859767.1
XM_017004279.1 Intron XP_016859768.1
XM_017004280.1 Intron XP_016859769.1
XM_017004281.1 Intron XP_016859770.1
XM_017004282.1 Intron XP_016859771.1
XM_017004283.1 Intron XP_016859772.1
XM_017004284.1 Intron XP_016859773.1
XM_017004285.1 Intron XP_016859774.1
XM_017004286.1 Intron XP_016859775.1
XM_017004287.1 Intron XP_016859776.1
XM_017004288.1 Intron XP_016859777.1
XM_017004289.1 Intron XP_016859778.1
XM_017004290.1 Intron XP_016859779.1
XM_017004291.1 Intron XP_016859780.1
Gene
GPC1
Gene Name
glypican 1
There are no transcripts associated with this gene.

View Full Product Details