Product Details

SNP ID
rs73196126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195520030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTATTTTTTCTTTTTTTCAA[A/G]ATGGAGTTTCACTCTAATTGCCCAG
Phenotype
MIM: 601792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R2 PubMed Links

Gene Details

Gene
PPP1R2
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291504.1 Intron NP_001278433.1
NM_001291505.1 Intron NP_001278434.1
NM_001316325.1 Intron NP_001303254.1
NM_006241.7 Intron NP_006232.1
XM_006713682.3 Intron XP_006713745.1

View Full Product Details