Product Details

SNP ID
rs73857319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113003676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGCCCAGTCCATGCTATATT[A/G]TCCTTAACTCACTGAACCACCCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
There are no transcripts associated with this gene.

Gene
NEPRO
Gene Name
nucleolus and neural progenitor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319109.1 3625 UTR 3 NP_001306038.1
NM_001319110.1 3625 UTR 3 NP_001306039.1
NM_001319111.1 3625 UTR 3 NP_001306040.1
NM_001319112.1 3625 UTR 3 NP_001306041.1
NM_001319114.1 3625 UTR 3 NP_001306043.1
NM_001319115.1 3625 UTR 3 NP_001306044.1
NM_015412.3 3625 UTR 3 NP_056227.2
XM_005247276.2 3625 Intron XP_005247333.1
XM_011512635.2 3625 Intron XP_011510937.1
XM_011512636.2 3625 Intron XP_011510938.1
XM_011512638.2 3625 Intron XP_011510940.1
XM_017006102.1 3625 UTR 3 XP_016861591.1
XM_017006103.1 3625 Intron XP_016861592.1

View Full Product Details