Product Details

SNP ID
rs73846074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93980167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCGACGCGGGGCTTTTCTTTAG[C/T]CGGGTCCCGCTAACTCGGCTACGGT
Phenotype
MIM: 608922 MIM: 176880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL13B PubMed Links

Gene Details

Gene
ARL13B
Gene Name
ADP ribosylation factor like GTPase 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174150.1 2 UTR 5 NP_001167621.1
NM_001174151.1 2 UTR 5 NP_001167622.1
NM_001321328.1 2 UTR 5 NP_001308257.1
NM_144996.3 2 UTR 5 NP_659433.2
NM_182896.2 2 UTR 5 NP_878899.1
XM_006713532.3 2 UTR 5 XP_006713595.1
XM_011512532.2 2 UTR 5 XP_011510834.1
XM_011512533.2 2 Intron XP_011510835.1
XM_011512534.2 2 UTR 5 XP_011510836.1
XM_011512535.2 2 UTR 5 XP_011510837.1
XM_017005853.1 2 UTR 5 XP_016861342.1
Gene
PROS1
Gene Name
protein S (alpha)
There are no transcripts associated with this gene.

View Full Product Details