Product Details

SNP ID
rs72954680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113953108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTATGTAGCCCTGTCCCTGTGG[G/T]ACTAGTCTACTCCCTGGGCTCATGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRAMD1C PubMed Links

Gene Details

Gene
GRAMD1C
Gene Name
GRAM domain containing 1C
There are no transcripts associated with this gene.

Gene
ZDHHC23
Gene Name
zinc finger DHHC-type containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320466.1 Intron NP_001307395.1
NM_001320467.1 Intron NP_001307396.1
NM_001320468.1 Intron NP_001307397.1
NM_173570.4 Intron NP_775841.2
XM_005247269.1 Intron XP_005247326.1
XM_005247270.3 Intron XP_005247327.1
XM_006713562.3 Intron XP_006713625.1
XM_011512618.2 Intron XP_011510920.1
XM_011512619.1 Intron XP_011510921.1
XM_017006084.1 Intron XP_016861573.1
XM_017006085.1 Intron XP_016861574.1
XM_017006086.1 Intron XP_016861575.1
XM_017006087.1 Intron XP_016861576.1

View Full Product Details