Product Details

SNP ID
rs72986627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146519018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAGGATCTAGGTATGTAGCAAA[G/T]AAATATAAAAAGTGTATCATGAGTA
Phenotype
MIM: 604170
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLSCR1 PubMed Links

Gene Details

Gene
PLSCR1
Gene Name
phospholipid scramblase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021105.2 Intron NP_066928.1
XM_005247538.3 Intron XP_005247595.1
XM_011512904.1 Intron XP_011511206.1
XM_011512905.1 Intron XP_011511207.1
XM_011512906.2 Intron XP_011511208.1
XM_011512907.2 Intron XP_011511209.1
XM_017006628.1 Intron XP_016862117.1
XM_017006629.1 Intron XP_016862118.1

View Full Product Details