Product Details

SNP ID
rs73058079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183184398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCCAGCCAAGATCTAGTGGT[A/G]TGTTGGAGCAGATACACCCTGAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCF2L2 PubMed Links

Gene Details

Gene
MCF2L2
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015078.3 Intron NP_055893.3
XM_011512585.2 Intron XP_011510887.1
XM_011512586.2 Intron XP_011510888.1
XM_017005943.1 Intron XP_016861432.1
XM_017005944.1 Intron XP_016861433.1
XM_017005945.1 Intron XP_016861434.1
XM_017005946.1 Intron XP_016861435.1

View Full Product Details