Product Details

SNP ID
rs73886536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187724161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCATCACCACCTAGAAGAATGT[C/T]ATCAACTCCTCAAAGCAACCATAGG
Phenotype
MIM: 109565 MIM: 609138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL6 PubMed Links

Gene Details

Gene
BCL6
Gene Name
B-cell CLL/lymphoma 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130845.1 Intron NP_001124317.1
NM_001134738.1 Intron NP_001128210.1
NM_001706.4 Intron NP_001697.2
XM_005247694.3 Intron XP_005247751.1
XM_011513062.2 Intron XP_011511364.1
Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
There are no transcripts associated with this gene.

View Full Product Details