Product Details

SNP ID
rs73199948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6918507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTTGCCAATGCCTTCAGCAGA[C/T]CTTCTCATTTCTTTTGTCCTCAGAC
Phenotype
MIM: 614855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D14 PubMed Links

Gene Details

Gene
TBC1D14
Gene Name
TBC1 domain family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113361.1 Intron NP_001106832.1
NM_001113363.1 Intron NP_001106834.1
NM_001286805.1 Intron NP_001273734.1
NM_020773.2 Intron NP_065824.2
XM_005247986.2 Intron XP_005248043.1
XM_006713895.2 Intron XP_006713958.1
XM_011513507.2 Intron XP_011511809.1
XM_011513508.1 Intron XP_011511810.1
XM_011513513.2 Intron XP_011511815.1
XM_017008477.1 Intron XP_016863966.1
XM_017008478.1 Intron XP_016863967.1
XM_017008479.1 Intron XP_016863968.1
XM_017008480.1 Intron XP_016863969.1

View Full Product Details