Product Details

SNP ID
rs73144650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48490529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGAGTACTTCGACAGCTACCAG[A/C]GGGAGCGGCTCATGGCTCTCCTGGC
Phenotype
MIM: 607520
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
There are no transcripts associated with this gene.

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
There are no transcripts associated with this gene.

Gene
ZAR1
Gene Name
zygote arrest 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175619.2 278 Silent Mutation AGG,CGG R80R NP_783318.1

View Full Product Details