Product Details

SNP ID
rs72839479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27365131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGATTAGTTTTCTCCCTCAAAGC[A/G]TTTTTAGGATACTTTTGTTTTGAAT
Phenotype
MIM: 603282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF204P PubMed Links

Gene Details

Gene
ZNF204P
Gene Name
zinc finger protein 204, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF391
Gene Name
zinc finger protein 391
There are no transcripts associated with this gene.

View Full Product Details