Product Details

SNP ID
rs70953697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43305487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAAGGCATGGGAGCCAACATTT[C/T]ATTGAAGAAGCCACAGAGGCTGAAA
Phenotype
MIM: 604995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRIP3 PubMed Links

Gene Details

Gene
CRIP3
Gene Name
cysteine rich protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206922.2 942 UTR 3 NP_996805.2
XM_005249103.3 942 Intron XP_005249160.1
XM_011514609.2 942 Intron XP_011512911.1
XM_017010856.1 942 Intron XP_016866345.1
XM_017010857.1 942 Intron XP_016866346.1
Gene
SLC22A7
Gene Name
solute carrier family 22 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006672.3 942 UTR 3 NP_006663.2
NM_153320.2 942 UTR 3 NP_696961.2
XM_006714970.3 942 UTR 3 XP_006715033.1
XM_006714971.3 942 UTR 3 XP_006715034.1
XM_011514256.2 942 UTR 3 XP_011512558.1
XM_011514257.2 942 UTR 3 XP_011512559.1
XM_011514259.1 942 Intron XP_011512561.1
XM_011514261.2 942 UTR 3 XP_011512563.1
XM_011514262.2 942 Intron XP_011512564.1
XM_011514263.2 942 UTR 3 XP_011512565.1
XM_017010198.1 942 UTR 3 XP_016865687.1
XM_017010199.1 942 UTR 3 XP_016865688.1
XM_017010200.1 942 UTR 3 XP_016865689.1
XM_017010201.1 942 UTR 3 XP_016865690.1

View Full Product Details