Product Details

SNP ID
rs73374668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66077091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATTGCCCACCTCATAAAAGGC[C/T]GTAATAACATATGTGAAATCCCTAG
Phenotype
MIM: 608310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASL PubMed Links

Gene Details

Gene
ASL
Gene Name
argininosuccinate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000048.3 Intron NP_000039.2
NM_001024943.1 Intron NP_001020114.1
NM_001024944.1 Intron NP_001020115.1
NM_001024946.1 Intron NP_001020117.1

View Full Product Details