Product Details

SNP ID
rs73097443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36514818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGGCAGCGTAGCCAACTCCCT[C/T]TCTGGGCCCAGCCCTCTCATCTGGA
Phenotype
MIM: 102593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AOAH PubMed Links

Gene Details

Gene
AOAH
Gene Name
acyloxyacyl hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177506.1 Intron NP_001170977.1
NM_001177507.1 Intron NP_001170978.1
NM_001637.3 Intron NP_001628.1
XM_011515333.2 Intron XP_011513635.1
XM_011515334.2 Intron XP_011513636.1
XM_011515335.2 Intron XP_011513637.1
XM_011515336.2 Intron XP_011513638.1
XM_011515338.2 Intron XP_011513640.1
XM_011515339.2 Intron XP_011513641.1
XM_011515340.2 Intron XP_011513642.1
XM_011515341.2 Intron XP_011513643.1
XM_011515342.2 Intron XP_011513644.1
XM_017012102.1 Intron XP_016867591.1
XM_017012103.1 Intron XP_016867592.1
XM_017012104.1 Intron XP_016867593.1
XM_017012105.1 Intron XP_016867594.1
XM_017012106.1 Intron XP_016867595.1

View Full Product Details