Product Details

SNP ID
rs73674126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6502086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGGCAAGGATGTGCAGGAAGGT[A/G]GACTTCCCATCCACTGTCTTGGTGG
Phenotype
MIM: 610639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRID2IP PubMed Links

Gene Details

Gene
GRID2IP
Gene Name
Grid2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145118.1 3183 Silent Mutation TCC,TCT S1061S NP_001138590.1
XM_011515404.2 3183 Silent Mutation TCC,TCT S877S XP_011513706.1

View Full Product Details