Product Details

SNP ID
rs73170678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143139110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCATTAGCAGCATCCCTCTACA[A/T]GGTGCATTTAACTATAAGTATACTG
Phenotype
MIM: 176720
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PIP PubMed Links

Gene Details

Gene
PIP
Gene Name
prolactin induced protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002652.2 273 Missense Mutation CAA,CAT Q79H NP_002643.1

View Full Product Details