Product Details

SNP ID
rs73713088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:104333796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTACAGCACATTTCCATGTTT[A/G]TTTAATTTCTGTCTTCTCCAGTGCA
Phenotype
MIM: 609719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFPL3 PubMed Links

Gene Details

Gene
LHFPL3
Gene Name
lipoma HMGIC fusion partner-like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199000.2 Intron NP_945351.1
XM_005250327.1 Intron XP_005250384.1

View Full Product Details