Product Details

SNP ID
rs73195075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112826957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGTTTATCAGTTTTTGGTTTA[C/T]ATATTTGGAAGCTTTGTTATTAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMT2 PubMed Links

Gene Details

Gene
BMT2
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152556.2 Intron NP_689769.2

View Full Product Details