Product Details

SNP ID
rs73634664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42840290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACATTTGTATCTGCGTACTGG[A/G]AAAAAGGGGCTTCTAGTTCTCTTAG
Phenotype
MIM: 614649 MIM: 609520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF170 PubMed Links

Gene Details

Gene
RNF170
Gene Name
ring finger protein 170
There are no transcripts associated with this gene.

Gene
THAP1
Gene Name
THAP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018105.2 Intron NP_060575.1
NM_199003.1 Intron NP_945354.1

View Full Product Details