Product Details

SNP ID
rs73669623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30105919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGTCATTTGTTGGCCATTCAC[A/G]CACACAGGAGATGGGGCAGTTAATG
Phenotype
MIM: 607338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LEPROTL1 PubMed Links

Gene Details

Gene
LEPROTL1
Gene Name
leptin receptor overlapping transcript-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128208.1 560 Intron NP_001121680.1
NM_015344.2 560 UTR 3 NP_056159.2

View Full Product Details