Product Details

SNP ID
rs73698505
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96143852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAAGGTTGCCAGTTCCTGGTA[A/C]CGAAAGAGGCCAGGGCAGAGCTGGT
Phenotype
MIM: 601147
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GDF6 PubMed Links

Gene Details

Gene
GDF6
Gene Name
growth differentiation factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001557.2 2179 UTR 3 NP_001001557.1

View Full Product Details