Product Details

SNP ID
rs72718565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:127800786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAAGAAAAAGGACAGATTTCC[A/G]TGCTAGAGCTTAGGTTACAGCTGGG
Phenotype
MIM: 165140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1204 PubMed Links

Gene Details

Gene
MIR1204
Gene Name
microRNA 1204
There are no transcripts associated with this gene.

Gene
PVT1
Gene Name
Pvt1 oncogene (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details