Product Details

SNP ID
rs73254833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:68331730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACACTTTCTTTTTTGAATTCAC[C/G]CTATCTTGAGTGTTCGTGTTGGATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C8orf34 PubMed Links

Gene Details

Gene
C8orf34
Gene Name
chromosome 8 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195639.1 Intron NP_001182568.1
NM_052958.2 Intron NP_443190.2
XM_011517445.2 Intron XP_011515747.1
XM_011517446.2 Intron XP_011515748.1
XM_011517448.2 Intron XP_011515750.1
XM_011517449.2 Intron XP_011515751.1
XM_011517450.2 Intron XP_011515752.1
XM_017013019.1 Intron XP_016868508.1
Gene
C8orf34-AS1
Gene Name
C8orf34 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details