Product Details

SNP ID
rs73691134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80041023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTAGGACTGTGTGTGTACAGT[A/G]TAACTCCTCTTCTGAAAAAACATTC
Phenotype
MIM: 604068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPD52 PubMed Links

Gene Details

Gene
TPD52
Gene Name
tumor protein D52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025252.2 Intron NP_001020423.1
NM_001025253.2 Intron NP_001020424.1
NM_001287140.1 Intron NP_001274069.1
NM_001287142.1 Intron NP_001274071.1
NM_001287143.1 Intron NP_001274072.1
NM_001287144.1 Intron NP_001274073.1
NM_005079.3 Intron NP_005070.1

View Full Product Details