Product Details

SNP ID
rs73679602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54049050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACATTAAGGGTTCCAGGACTTC[C/T]CCTGCCCTCTCTTTTCTGGATTCAA
Phenotype
MIM: 605599
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105375839 PubMed Links

Gene Details

Gene
LOC105375839
Gene Name
uncharacterized LOC105375839
There are no transcripts associated with this gene.

Gene
LYPLA1
Gene Name
lysophospholipase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001279356.1 Intron NP_001266285.1
NM_001279357.1 Intron NP_001266286.1
NM_001279358.1 Intron NP_001266287.1
NM_001279359.1 Intron NP_001266288.1
NM_001279360.1 Intron NP_001266289.1
NM_006330.3 Intron NP_006321.1
XM_005251127.4 Intron XP_005251184.1
XM_017012956.1 Intron XP_016868445.1
XM_017012957.1 Intron XP_016868446.1
XM_017012958.1 Intron XP_016868447.1
XM_017012959.1 Intron XP_016868448.1
XM_017012960.1 Intron XP_016868449.1

View Full Product Details