Product Details

SNP ID
rs73602288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56380358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTTTCTTGGCACATAATTTTTC[A/G]TAGTATTTCCTTATGGTCCTTTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDR16C6P PubMed Links

Gene Details

Gene
SDR16C6P
Gene Name
short chain dehydrogenase/reductase family 16C, member 6, pseudogene
There are no transcripts associated with this gene.

View Full Product Details