Product Details

SNP ID
rs73517867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104783899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCATGGCATGGCTTAGTGAAT[A/G]AGGATGTGCATTACCTTTTGATTTT
Phenotype
MIM: 600046 MIM: 608872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA1 PubMed Links

Gene Details

Gene
ABCA1
Gene Name
ATP binding cassette subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005502.3 7521 UTR 3 NP_005493.2
XM_005251773.2 7521 UTR 3 XP_005251830.1
XM_005251776.2 7521 UTR 3 XP_005251833.1
XM_011518339.2 7521 UTR 3 XP_011516641.1
XM_011518340.2 7521 UTR 3 XP_011516642.1
XM_011518341.2 7521 UTR 3 XP_011516643.1
XM_011518342.2 7521 UTR 3 XP_011516644.1
XM_011518344.2 7521 Intron XP_011516646.1
XM_017014378.1 7521 UTR 3 XP_016869867.1
XM_017014379.1 7521 UTR 3 XP_016869868.1
XM_017014380.1 7521 UTR 3 XP_016869869.1
XM_017014381.1 7521 UTR 3 XP_016869870.1
XM_017014382.1 7521 UTR 3 XP_016869871.1
Gene
NIPSNAP3B
Gene Name
nipsnap homolog 3B
There are no transcripts associated with this gene.

View Full Product Details