Product Details
- SNP ID
-
rs73663057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:122240256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCCAAGAAGTCACCAGAAAAAT[A/G]GGAGATAATGCTCCTACCTCTCTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBM18
PubMed Links
Gene Details
- Gene
- RBM18
- Gene Name
- RNA binding motif protein 18
View Full Product Details