Product Details

SNP ID
rs73565661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122583177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGAGTAAACAATAAATGGATAC[A/G]CCCCTATTCACTCTAGTCTGATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1B1 PubMed Links

Gene Details

Gene
OR1B1
Gene Name
olfactory receptor family 1 subfamily B member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004450.1 Intron NP_001004450.1
XM_017014695.1 Intron XP_016870184.1
XM_017014696.1 Intron XP_016870185.1
Gene
OR1L8
Gene Name
olfactory receptor family 1 subfamily L member 8
There are no transcripts associated with this gene.

View Full Product Details