Product Details
- SNP ID
-
rs72616651
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:124353890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCTCAGTTCTCAAGTGCTGCAA[A/G]ATGGGGGTGCCTGGGAGGCAAAGAG
- Phenotype
-
MIM: 604884
MIM: 604030
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100129034
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144525600] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100129034
- Gene Name
- uncharacterized LOC100129034
There are no transcripts associated with this gene.
- Gene
- NEK6
- Gene Name
- NIMA related kinase 6
There are no transcripts associated with this gene.
- Gene
- PSMB7
- Gene Name
- proteasome subunit beta 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002799.3 |
|
Intron |
|
|
NP_002790.1 |
View Full Product Details