Product Details

SNP ID
rs72743247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94559122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGCCACGGGATTGCATTCATAC[A/G]GGAGCCGGAGCTGTGGAGGAACAGA
Phenotype
MIM: 603027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBP2 PubMed Links

Gene Details

Gene
FBP2
Gene Name
fructose-bisphosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003837.3 942 Missense Mutation CCG,CTG P279L NP_003828.2
Gene
PCAT7
Gene Name
prostate cancer associated transcript 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details