Product Details

SNP ID
rs74144723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79384881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAGAGACCCAGTTCTGACAGC[C/T]GGGAGAAAGGAAGGGTCAAGACCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC24 PubMed Links

Gene Details

Gene
ZCCHC24
Gene Name
zinc finger CCHC-type containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153367.3 2328 UTR 3 NP_699198.2
XM_011539452.2 2328 UTR 3 XP_011537754.1

View Full Product Details