Product Details

SNP ID
rs72843154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127739557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCATTGGGTCCTGTGTAAGGAA[A/G]GTCTTGCTAATGGTTGGAACTGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXI2 PubMed Links

Gene Details

Gene
FOXI2
Gene Name
forkhead box I2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207426.2 1549 UTR 3 NP_997309.2
Gene
LOC387720
Gene Name
collagen alpha-1(X) chain
There are no transcripts associated with this gene.

View Full Product Details