Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001313726.1 | 5065 | Intron | NP_001300655.1 | ||
NM_001313727.1 | 5065 | Intron | NP_001300656.1 | ||
NM_031418.3 | 5065 | Intron | NP_113606.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_178498.3 | 5065 | UTR 3 | NP_848593.2 | ||
XM_006718155.3 | 5065 | UTR 3 | XP_006718218.1 | ||
XM_006718156.3 | 5065 | Intron | XP_006718219.1 | ||
XM_011519920.2 | 5065 | UTR 3 | XP_011518222.1 | ||
XM_011519921.2 | 5065 | Intron | XP_011518223.1 | ||
XM_017017244.1 | 5065 | UTR 3 | XP_016872733.1 |