Product Details

SNP ID
rs72888131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31264485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAACAGAGTACAATGGTTAAAC[A/G]CTGCTACTGGAGCCAGCCTGCCAAG
Phenotype
MIM: 608062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCDC1 PubMed Links

Gene Details

Gene
DCDC1
Gene Name
doublecortin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181807.3 Intron NP_861523.2

View Full Product Details