Product Details

SNP ID
rs72901247
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32917389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATGGGAGTAGAGTTGCTGGGCC[A/G]TGTGGTAATGATGTTTAACTTTTTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
QSER1 PubMed Links

Gene Details

Gene
QSER1
Gene Name
glutamine and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076786.2 Intron NP_001070254.1
XM_006718325.3 Intron XP_006718388.3
XM_017018327.1 Intron XP_016873816.1
XM_017018328.1 Intron XP_016873817.1
XM_017018329.1 Intron XP_016873818.1
XM_017018330.1 Intron XP_016873819.1

View Full Product Details