Product Details
- SNP ID
-
rs73052536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:7694618 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAACTTTATCTTGTATTCTCAAG[A/G]CCATACACAGTTCCTGGCTCACCAT
- Phenotype
-
MIM: 606522
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GDF3
PubMed Links
Gene Details
- Gene
- GDF3
- Gene Name
- growth differentiation factor 3
There are no transcripts associated with this gene.
View Full Product Details