Product Details

SNP ID
rs73032663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1624417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAGAGAAAGCAGAGAAG[A/G]TGTTATGCTCTAGTGACAGGGAGGA
Phenotype
MIM: 606361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT5B PubMed Links

Gene Details

Gene
WNT5B
Gene Name
Wnt family member 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030775.2 Intron NP_110402.2
NM_032642.2 Intron NP_116031.1

View Full Product Details