Product Details

SNP ID
rs71583750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4273820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTTTAAGGGGAGGACCGGTGC[A/G]AGTGAGGCAGCCCCGAGGCTCTGCT
Phenotype
MIM: 123833
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCND2 PubMed Links

Gene Details

Gene
CCND2
Gene Name
cyclin D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001759.3 85 UTR 5 NP_001750.1
Gene
CCND2-AS1
Gene Name
CCND2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details