Product Details

SNP ID
rs71583765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4434279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACCTTGCTGCCCCGCTTTACC[C/T]GTCCGTATTTGCTCAGGGCAATGTA
Phenotype
MIM: 134921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF6 PubMed Links

Gene Details

Gene
FGF6
Gene Name
fibroblast growth factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020996.2 607 Missense Mutation NP_066276.2
XM_017018995.1 607 Intron XP_016874484.1
XM_017018996.1 607 Intron XP_016874485.1

View Full Product Details