Product Details

SNP ID
rs73266929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6696040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGACACCTGAGTCTAATGATTCT[C/G]TCAGTAAAGTGTGAAGGACTGAGTG
Phenotype
MIM: 616065 MIM: 609951
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PIANP PubMed Links

Gene Details

Gene
PIANP
Gene Name
PILR alpha associated neural protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244014.1 Intron NP_001230943.1
NM_001244015.1 Intron NP_001230944.1
NM_153685.3 Intron NP_710152.1
XM_011520926.2 Intron XP_011519228.1
Gene
ZNF384
Gene Name
zinc finger protein 384
There are no transcripts associated with this gene.

View Full Product Details