Product Details

SNP ID
rs73412044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:119180086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGCACGTTAAGGCTGAACAATG[A/G]TAGTGTTAATAACCGCAACTGGTTT
Phenotype
MIM: 608014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPB8 PubMed Links

Gene Details

Gene
HSPB8
Gene Name
heat shock protein family B (small) member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014365.2 Intron NP_055180.1
Gene
LOC105370024
Gene Name
uncharacterized LOC105370024
There are no transcripts associated with this gene.

View Full Product Details