Product Details

SNP ID
rs73409738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121628134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTTGGCATATTGAAGTTTCTC[A/C]GTTGCAGGGGACTACATAATAACTA
Phenotype
MIM: 610277
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ORAI1 PubMed Links

Gene Details

Gene
ORAI1
Gene Name
ORAI calcium release-activated calcium modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032790.3 Intron NP_116179.2

View Full Product Details